Learn More
Cayman Chemical Amodaqun 10mg
SDP
Supplier:  Cayman Chemical 1595410 MG
Encompass_Preferred
A prodrug form of N-desethyl amodiaquine; active against several strains of P. falciparum in vitro (EC50s = 0.23-0.52 nM) and exhibits a synergistic effect when used in combination with N-desethyl amodiaquine; dose-dependently inhibits development of parasitemia in a mouse model of P. berghei infection
Catalog No. 50-297-1605