Learn More
TARGETMOL CHEMICALS INC Amodiaquine dihydrochloride di
SDP
Supplier:  TARGETMOL CHEMICALS INC T0381500MG
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Amodiaquine dihydrochloride dihydrate (Amodiaquine hydrochloride) is the hydrochloride salt of amodiaquine, an orally active 4-aminoquinoline derivative with antimalarial and anti-inflammatory properties. Purity 99.9%
Catalog No. 50-288-0690