Learn More
Medchemexpress LLC Amodiaquine impurity 1 | 81099-86-7 | 270.72 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0087275G
Encompass_Preferred
Amodiaquine impurity 1 is a chemical compound identified as an impurity of Amodiaquine. It is intended for laboratory use as a drug intermediate in the manufacture of substances.
  • Suitable for laboratory chemical applications
  • Recommended storage at 4°C, protected from light and stored under nitrogen
  • Stable in solvent for up to 6 months at -80°C or 1 month at -20°C, protected from light and stored under nitrogen
Catalog No. 50-003-66341