Learn More
Medchemexpress LLC Amodiaquine impurity 1 | 81099-86-7 | 98.7% | 270.72 | 1 G

Supplier: Medchemexpress LLC HYW0087271G
Amodiaquine impurity 1 is a chemical compound with the molecular formula C15H11ClN2O, appearing as a light yellow to yellow solid. It is intended for use as a laboratory chemical and in the manufacture of substances.
- High purity of 98.70% by HPLC
- Molecular formula: C15H11ClN2O
- Appearance: light yellow to yellow solid
- Stable under recommended storage conditions
- Requires protection from light and storage under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.