Learn More
Medchemexpress LLC Amodiaquine impurity 1 | 81099-86-7 | 98.7% | 270.72 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW008727100MG
Encompass_Preferred
Amodiaquine impurity 1 is a chemical substance used in laboratory research and the manufacture of other substances. This compound is identified as an impurity of Amodiaquine and typically appears as a solid.
  • Used for laboratory research and manufacturing purposes
  • Supplied as a solid
  • Recommended storage at 4°C, protected from light, and under nitrogen
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C, protected from light and under nitrogen
Catalog No. 50-004-51170