Learn More
Selleck Chemical LLC Amonafide 10mg 69408-81-7 NSC308847,AS1413
SDP
Supplier:  Selleck Chemical LLC S136710MG
Encompass_Preferred
Amonafide (NSC308847, AS1413) produces protein-associated DNA-strand breaks through a topoisomerase II-mediated reaction, but does not produce topoisomerase I-mediated DNA cleavage. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-786-4