Learn More
TARGETMOL CHEMICALS INC Amorolfine hydrochloride 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0187200MG
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Amorolfine hydrochloride (Amorolfine HCl) is an antifungal reagent. It exerts the antifungal activity by selectively interrupting two steps in the pathway of ergosterol synthesis and eventually disrupting the function and structure of fungal cell membrane. Amorolfine hydrochloride , a morpholine antifungal drug, can inhibit D14 reductase and D7-D8 isomerase. These enzymes can deplete ergosterol and cause ignosterol to accumulate in the fungal cytoplasmic cell membranes. Purity 100%
Catalog No. 50-288-0671