Learn More
Medchemexpress LLC HY-107004A 5mg , Amotosalen (hydrochloride) CAS:161262-45-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107004A5MG
Encompass_Preferred
HY-107004A 5mg Amotosalen (hydrochloride) CAS: 161262-45-9 Amotosalen hydrochloride (S-59) is a light-activated, DNA-, RNA-crosslinking psoralen compound, which is used to neutralise pathogens. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1265