Learn More
Cayman Chemical AmoxIcIlIn-d4 1mg
SDP
Supplier:  Cayman Chemical 254281 MG
Encompass_Preferred
An internal standard for the quantification of amoxicillin by GC- or LC-MS
Catalog No. 50-293-5932