Learn More
Apexbio Technology LLC Amoxicillin 26787-78-0 5g
SDP
Supplier:  Apexbio Technology LLC B16615000
Encompass_Preferred
Amoxicillin (26787-78-0) is a small-molecule inhibitor targeting bacterial penicillin-binding proteins (PBPs) It is designed to disrupt bacterial cell wall biosynthesis thereby impairing peptidoglycan cross-linking and compromising cell wall integrity Amoxicillin exerts its biological activity primarily through inhibition of PBPs interfering with the processes critical to bacterial cell wall formation In in vitro studies Amoxicillin demonstrates inhibitory activity with IC50 values ranging from 0 1 to 5 g/mL depending on experimental conditions and organism susceptibility Based on these pharmacological properties Amoxicillin holds research potential in studies of bacterial infection mechanisms antimicrobial responses and resistance development
Catalog No. 50-000-92487