Learn More
Medchemexpress LLC Amoxicillin | 26787-78-0 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0467A10MM/1ML
Encompass_Preferred
Amoxicillin is an antibiotic characterized by good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the cell wall, which in turn inhibits cell growth. This product is intended for research use only.
  • Antibiotic with good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits biosynthesis of polypeptides in the cell wall
  • Inhibits cell growth
  • For research use only
Catalog No. 50-004-07804