Learn More
Medchemexpress LLC Amoxicillin | 26787-78-0 | 99.9% | 365.40 | 500 MG
SDP
Supplier:  Medchemexpress LLC 500MGHYB0467A
Encompass_Preferred
Amoxicillin is an antibiotic characterized by good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the cell wall, which in turn inhibits bacterial cell growth.
  • Broad-spectrum antimicrobial activity
  • Inhibits biosynthesis of polypeptides in the cell wall
  • Good oral absorption
  • For research use only
Catalog No. 50-003-65878