Learn More
Medchemexpress LLC Amoxicillin (Amoxycillin) | 26787-78-0 | >99.9% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB0467A100G
Encompass_Preferred
Amoxicillin (Amoxycillin) is an antibiotic characterized by good oral absorption and a broad spectrum of antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the cell wall, which in turn hinders cell growth.
  • Good oral absorption
  • Broad spectrum antimicrobial activity
  • Inhibits biosynthesis of polypeptides in the cell wall
  • Inhibits cell growth
Catalog No. 50-004-06770