Learn More
Medchemexpress LLC Amoxicillin-clavulanate potassium | 74469-00-4 | MFCD11041067 | 93.7% | C24H27KN4O10S | 10MG

Supplier: Medchemexpress LLC HY13553210MG
Amoxicillin-clavulanate potassium is a combination antibiotic (amoxicillin with clavulanate as the potassium salt) supplied for research use. The product is provided as a solid and as a DMSO solution for in vitro studies, and documentation includes chemical identifiers, solubility data, purity, and handling recommendations.
- Combination antibiotic used in bacterial infection and β-lactamase research.
- Available as solid and as 10 mM solution in DMSO.
- Chemical formula C24H27KN4O10S; molecular weight 602.66 g/mol.
- Soluble in DMSO (≥ 200 mg/mL); heat or sonication may aid dissolution.
- Purity 93.7% per certificate of analysis.
- Storage and handling guidance provided in the certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.