Learn More
Medchemexpress LLC Amoxicillin (sodium) | 34642-77-8 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB04675G
Encompass_Preferred
Amoxicillin (sodium) is an antibiotic with good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth. This product is intended for research use only.
  • Inhibits biosynthesis of polypeptides in the cell wall
  • Decreases living cells and increases cell wall rupture in L. acidophilus (in vitro)
  • Inhibits bacterial numbers in mice (in vivo)
  • Improves activity against Chlamydia trachomatis infection (in vivo)
  • Supporting documentation available, including data sheets, COA, and SDS
Catalog No. 50-004-08447