Learn More
Medchemexpress LLC Amoxicillin sodium | 34642-77-8 | MFCD08063910 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB046710MM/1ML
Encompass_Preferred
Amoxicillin sodium is an antibiotic with good oral absorption and broad-spectrum antimicrobial activity. It inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.
  • Good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits biosynthesis of polypeptides in the cell wall
  • Inhibits cell growth
Catalog No. 50-004-08219