Learn More
Alkali Scientific Amoxicillin Trihydrate, 10 G
SDP
Supplier:  Alkali Scientific AMO001CP
Encompass_Preferred
Amoxicillin Trihydrate is a broad spectrum penicillin class antibiotic widely used in microbiological and biochemical research. It works by inhibiting bacterial cell wall synthesis, leading to cell lysis and death. This compound is effective against a range of Gram positive and certain Gram negative bacteria and is frequently used in laboratory studies involving antimicrobial activity and bacterial growth control. In cell culture applications, it may be used to reduce bacterial contamination. Amoxicillin is also used in research focused on antibiotic resistance, pharmacological testing, and microbial inhibition, providing consistent and well characterized antibacterial performance.
Catalog No. 50-006-60844