Learn More
Medchemexpress LLC Amoxicillin trihydrate | 61336-70-7 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0467B5G
Encompass_Preferred
Amoxicillin trihydrate is an antibiotic with good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the cell wall, thereby inhibiting bacterial cell growth. This product is intended for research use only.
  • Good oral absorption
  • Broad-spectrum antimicrobial activity
  • Inhibits bacterial cell wall synthesis
  • Decreases living cells and increases cell wall rupture in vitro
  • Inhibits bacterial numbers and improves survival rate in vivo
  • Effective against Chlamydia trachomatis infection in vivo
Catalog No. 50-004-08218