Learn More
Medchemexpress LLC Amoxicillin trihydrate mixture with potassium clavulanate (4:1) | 100 MG
SDP
Supplier:  Medchemexpress LLC HY131165100MG
Encompass_Preferred
Amoxicillin trihydrate mixture with potassium clavulanate (4:1) is an antibiotic with good oral absorption and broad-spectrum antimicrobial activity. It functions by inhibiting the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.
  • Decreases living cells and increases the degree of cell wall rupture in L. acidophilus in a dose-dependent manner
  • Inhibits bacterial numbers and improves survival rate in female ICR/Swiss mice infected with organisms having MICs of 1 mg/L or less
  • Shows activity against infection with Chlamydia trachomatis in swiss albino mice
Catalog No. 50-003-41140