Learn More
Medchemexpress LLC Amoxicillin (trihydrate) mixture with potassium clavulanate (4:1) | 61336-70-7 | ≥98% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13116550MG
Encompass_Preferred
Amoxicillin (trihydrate) mixture with potassium clavulanate (4:1) is an antibiotic with good oral absorption and broad-spectrum antimicrobial activity. It inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.
  • Good oral absorption.
  • Broad-spectrum antimicrobial activity.
  • Inhibits cell wall polypeptide biosynthesis.
  • Inhibits cell growth.
  • Decreases living cells and increases cell wall rupture (in vitro).
  • Inhibits bacterial numbers (in vivo).
  • Improves activity against *Chlamydia trachomatis* infection (in vivo).
Catalog No. 50-003-39825