Learn More
Medchemexpress LLC AMP-Deoxynojirimycin 50mg | 216758-20-2 | 397.55 | C22H39NO5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11461550MG
Encompass_Preferred
AMP-Deoxynojirimycin (AMP-DNM) is a small-molecule inhibitor of ceramide glucosyltransferase and non-lysosomal glucosylceramidase (GCase2). It is used as a biochemical tool to inhibit glucosylceramide (GlcCer) biosynthesis in cellular and animal studies.
  • Inhibits ceramide glucosyltransferase and GCase2, blocking GlcCer biosynthesis.
  • Suitable as a research tool for studying glycosphingolipid metabolism and related pathways.
  • Provided as a solid powder for preparation of assay solutions.
  • Available in multiple pack sizes, including 50 mg.
  • Reported purity ~98% (see manufacturer documentation).
Catalog No. 50-002-29998