Learn More
Medchemexpress LLC AMP-Deoxynojirimycin 5mg | 216758-20-2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1146155MG
Encompass_Preferred
AMP-Deoxynojirimycin (AMP-DNM) is a research-grade small molecule inhibitor of ceramide glucosyltransferase and non-lysosomal glucocerebrosidase (GBA2). It reduces glucosylceramide (GlcCer) biosynthesis and is used in cellular and animal studies to probe glycosphingolipid metabolism, inflammation, and metabolic disease. Supplied as a solid for research use only.
  • Inhibits ceramide glucosyltransferase and GCase2.
  • Reduces glucosylceramide (GlcCer) biosynthesis.
  • Used in cellular and animal models to study glycosphingolipid metabolism and inflammation.
  • CAS number 216758-20-2; molecular formula C22H39NO5; molecular weight 397.55 g/mol.
  • Batch purities reported up to 99.7% (HPLC), depending on lot.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-07579