Learn More
Sino Biological Assay Buffer and Co-factors: AMP Solution
Supplier:  Sino Biological A4609500
Encompass_Preferred
0.5mM AMP solution used for AMPK family kinase assays.
Catalog No. 50-256-9740