Learn More
ATCC Ampelomyces quisqualis Cesati, cryopreserved culture (strain G273) - frozen ampoule for research
Supplier:  ATCC 200245SPQ
Encompass
Ampelomyces quisqualis Cesati is a mycoparasitic fungus supplied as a cryopreserved culture for research into biological control of powdery mildew. The culture is intended for laboratory propagation, characterization, and agricultural research involving fungal antagonists.
  • Mycoparasitic fungus that antagonizes powdery mildew.
  • Strain: G273.
  • Isolated from powdery mildew (Uncinula necator) on Vitis interspecific hybrid 'Rosette' leaves.
  • Supplied frozen in an ampoule containing viable cells suspended in cryoprotectant.
  • Intended for research use in agricultural and biological control studies.
Catalog No. NC3788955