Learn More
Medchemexpress LLC Amperozide | 75558-90-6 | 98.8% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12130110MG
Encompass_Preferred
Amperozide is an atypical antipsychotic research compound that antagonizes 5-HT2 receptors (Ki = 26 nM) and shows low affinity for D2 receptors. It is used as a tool in preclinical receptor-binding and pharmacology studies.
  • Acts as a 5-HT2 receptor antagonist (Ki = 26 nM).
  • Low affinity for D2 receptors.
  • Molecular formula C23H29F2N3O and molecular weight 401.49.
  • Listed purity approximately 98.8%.
  • Supplied in small research quantities suitable for in vitro and preclinical studies.
Catalog No. 50-002-32214