Learn More
Cayman Chemical AmphoterIcIn B 1g
SDP
Supplier:  Cayman Chemical 116361 G
Encompass_Preferred
A polyene macrolide used in the treatment of infections caused by C. albicans, A. fumigatus, and parasitic L. protozoans, as well as in tissue culture to prevent fungi from contaminating cell cultures; binds with ergosterol in fungal cell membranes, altering membrane permeability and ultimately inducing cell death
Catalog No. 50-295-5905