Learn More
Selleck Chemical LLC Amphotericin B 50mg 1397-89-3 NSC 527017
SDP
Supplier:  Selleck Chemical LLC S163650MG
Encompass_Preferred
Amphotericin B (AMB, NSC 527017) is an amphipathic polyene antibiotic which permeabilizes ergosterol-containing membranes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1459