Learn More
Selleck Chemical LLC Amphotericin B S1636-1g
SDP
Supplier:  Selleck Chemical LLC S16361G
Encompass_Preferred
Amphotericin B (AMB NSC 527017) is an amphipathic polyene antibiotic which permeabilizes ergosterol-containing membranes
Catalog No. 50-311-0679