Learn More
Abcam Amphotericin B (AmB), Antimycotic polyene, 1G
Supplier:  Abcam AB1411991G
Encompass_Preferred
MW 924.1 Da, Purity >90%. Antimycotic polyene. Disrupts fungal cell membrane permeability by targeting sterols such as ergosterol. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6810