Learn More
Quality Biological Inc Amphotericin B/Pen/Strep Mix 50ML, (120-096-711EA) QBICF0079
SDP
Supplier:  Quality Biological Inc 120096711
Encompass_Preferred
Amphotericin B 250 ¿g/ml/Penicillin10,000 Units/ml/ Streptomycin 10,000 ¿g/ml, prepared with cell culture grade water, 0.1micron filtered and cytotoxicity tested
Catalog No. 50-983-228