Learn More
Cayman Chemical AmpIcIlInhydrate 25g
SDP
Supplier:  Cayman Chemical 3574025 G
Encompass_Preferred
A broad-spectrum antibiotic; active against Gram-positive and Gram-negative bacteria, including veterinary isolates of S. pseudintermedius, S. aureus, E. coli, Pasteurella, and S. canis (MIC50s = 0.25, 0.5, 2, 0.12, and 0.25 μg/ml, respectively); reduces cough frequency, tachypnea, dyspnea, and fever and increases survival in a baboon (P. cynocephalus) model of pneumococcal pneumonia at 80 mg/kg
Catalog No. 50-291-0405