Learn More
Zymo Research Corporation Ampicillin Sodium Solution, 5x5 ml
SDP
Supplier:  Zymo Research Corporation A100125
Encompass
Resistance to ampicillin is conferred by the bla gene which encodes β-lactamase that cleaves the β-lactam bond of the antibiotic. Ampicillin is commonly used to select for ampicillin resistant plasmid bearing strains of bacteria.
Catalog No. 50-195-7624