Learn More
Alkali Scientific Ampicillin, Sodium Salt, 1 Kg
SDP
Supplier:  Alkali Scientific MPI004CP
Encompass
Ampicillin Sodium Salt is a high purity broad spectrum beta lactam antibiotic supplied in bulk powder form for laboratory and industrial scale applications. It functions by inhibiting bacterial cell wall synthesis, making it effective against a wide range of Gram positive and Gram negative organisms. This product is commonly used in large volume culture preparation, fermentation processes, and molecular biology workflows that require selective growth conditions. The powder format allows flexible preparation of stock solutions at desired concentrations. It is suitable for microbiological assays, research studies, and routine laboratory operations where consistent antibacterial performance and reproducibility are essential.
Catalog No. 50-006-61635