Learn More
Medchemexpress LLC Ampicillin (sodium) | 69-52-3 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0522A1G
Encompass_Preferred
Ampicillin sodium is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria.
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Inhibits *E. coli* growth in a dose-dependent manner
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Reduces MMP activities and increases GLT-1 expression
Catalog No. 50-004-06871