Learn More
Alkali Scientific Ampicillin, Sodium Salt, 10 G
SDP
Supplier:  Alkali Scientific MPI002CP
Encompass_Preferred
Ampicillin Sodium Salt is a broad spectrum beta lactam antibiotic provided as a high purity powder for laboratory use. It functions by inhibiting bacterial cell wall synthesis, leading to effective suppression of susceptible Gram positive and Gram negative bacteria. This reagent is widely used in molecular biology for selective growth of recombinant organisms carrying ampicillin resistance markers. The powder form allows accurate preparation of stock solutions for culture media and experimental applications. It is suitable for microbiological testing, antibiotic susceptibility studies, and routine laboratory workflows requiring reliable antibacterial activity and consistent experimental performance.
Catalog No. 50-006-61656