Learn More
Bioworld AMPICILLIN SODIUM SALT
Supplier:  Bioworld 401100321
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. Synonyms : D-()--Aminobenzylpenicillinsodium salt A -lactam antibiotic with an amino group side chain attached to the penicillin structure, 6-amino-penicillanic acid. A semi-synthetic penicillin derivative that inhibits bacterial cell-wa...
Catalog No. 50-255-302