Learn More
AG Scientific Inc Ampicillin, Sodium Salt, 5 G
Supplier:  AG Scientific Inc A14145GM
Encompass_Preferred
Ampicillin Sodium Salt is a Beta-Lactam antibiotic within the penicillin family. It inhibits synthesis of bacterial cell wall by inactivation of transpeptidases. It is used as a selective agent of ampicillin resistance in mutated and transformed cells having plasmid encoding for Beta-Lactamase production such as pBR322 or pUC.It is sensitive to Beta-lactamase, which hydrolyzes Beta-lactam ring. This antibiotic is effective against gram positive and highly effective against gram negative bacteria.CAS Number: 69-52-3Molecular Weight: 371.4 DaChemical Formula: C16H18N3O4S NaSolubility: Soluble in waterStorage Temperature: +4CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6884