Learn More
Alkali Scientific Ampicillin Trihydrate, 25 G
SDP
Supplier:  Alkali Scientific PIC002CP
Encompass_Preferred
Ampicillin Trihydrate is a broad spectrum antibiotic provided as a high purity powder for laboratory use. It functions by inhibiting bacterial cell wall synthesis, effectively suppressing susceptible Gram positive and Gram negative organisms. This reagent is commonly used in selective culture media for growth of resistant strains and in molecular biology workflows involving recombinant organisms. The powder form allows accurate preparation of stock solutions tailored to specific experimental requirements. It is suitable for microbiological assays, antibacterial research, and routine laboratory procedures that require dependable and reproducible antibiotic activity.
Catalog No. 50-006-61264