Learn More
Abcam Ampicillin trihydrate, beta-lactam antibiotic, 5G
Supplier:  Abcam AB1412005G
Encompass_Preferred
MW 349.4 Da, Purity >98%. β-lactam antibiotic (MIC₅₀ = 3.12 μg/ml). Competitively inhibits transpeptidase causing cell wall disruption to Gram-positive and Gram-negative bacteria. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6807