Learn More
Enzo Life Sciences AMPIGENE cDNA Synthesis Kit (50 Reactions)
SDP
Supplier:  Enzo Life Sciences ENZKIT1060050
Encompass_Preferred
AMPIGENE cDNA Synthesis Kit uses the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient and accurate cDNA synthesis. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. An optimized buffer system contains oligo(dT), random hexamers, enhancers, dNTPs and MgCl2, allowing efficient and sensitive cDNA synthesis.
Catalog No. 50-201-3812