Learn More
Sino Biological Recombinant Human PLK1 Protein (Full Length, His Tag), Active
Supplier:  Sino Biological P4110H50
Encompass_Preferred
Recombinant full length human PLK1 was expressed by baculovirus in Sf9 insect cells using a N-terminal His tag.
Catalog No. 50-257-3681