Learn More
Medchemexpress LLC AMPK alpha 1 Antibody | 64 kDa | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8053950UL
Encompass_Preferred
The AMPK alpha 1 Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed to target AMPK alpha 1. This protein is a catalytic subunit of the 5'-prime-AMP-activated protein kinase (AMPK), a critical cellular energy sensor that helps protect cells from ATP depletion by regulating metabolic enzymes through phosphorylation.
  • Suitable for Western Blotting (WB) at 1:500-1:1000 dilution.
  • Suitable for Immunohistochemistry-Paraffin (IHC-P) at 1:50-1:100 dilution.
  • Suitable for Immunocytochemistry/Immunofluorescence (ICC/IF) at 1:50-1:200 dilution.
  • Suitable for Immunoprecipitation (IP) at 1:20 dilution.
  • Suitable for Flow Cytometry (FC) at 1:50-1:100 dilution.
Catalog No. 50-003-95795