Learn More
Medchemexpress LLC AMPK alpha 2 Antibody (YA4091) (PBS only) | 62 kDa | 100 UL
SDP
Supplier:  Medchemexpress LLC HYP84394100UL
Encompass_Preferred
AMPK alpha 2 Antibody (YA4091) is a mouse-derived IgG1 monoclonal antibody targeting AMPK alpha 2, a central energy sensor kinase that regulates cellular metabolism. It activates catabolic pathways and inhibits anabolic processes under ATP depletion, playing a crucial role in maintaining energy homeostasis. This antibody is supplied in PBS, pH 7.4.
  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets AMPK alpha 2, a key regulator of metabolism
  • Reacts with Human, Mouse, Rat, Monkey, and Rabbit samples
  • Suitable for WB, IHC-P, ICC/IF, FC, and ELISA applications
  • Affinity purified
Catalog No. 50-004-45429