Learn More
Medchemexpress LLC Ampk-in-3 | 2417674-27-0 | C25H33N5O3 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY15136110MM/1ML
Encompass_Preferred
AMPK-IN-3 is a potent and selective AMPK inhibitor with IC50s of 60.7 nM for AMPK (α2) and 107 nM for AMPK (α1). It is described as a yellow to brown solid, and it inhibits AMPK without affecting cell viability or causing significant cytotoxicity in K562 cells. This compound can be used in the study of cancer.
  • Potent and selective inhibitor of AMPK
  • Does not affect cell viability
  • Does not cause significant cytotoxicity in K562 cells
  • Useful in cancer research
  • Solid appearance, yellow to brown color
  • Soluble in DMSO
Catalog No. 50-003-72233