Learn More
Sino Biological Recombinant Human AMPK (G1/B2/A2) Heterotrimer Protein (Full Length, A2/B2/G1, His & GST Tag), Active
Supplier:  Sino Biological P4910GH50
Encompass_Preferred
Recombinant full length human AMPK (combination of A2/B2/G1 subunits) was expressed by baculovirus in Sf9 insect cells using the N-terminal GST and C-terminal His tags.
Catalog No. 50-257-3649