Learn More
Alkali Scientific AMPSO [3 ((1,1 Dimethyl 2 hydroxyethyl)amino) 2 hydroxypropanesulfonic acid], 25 G
Supplier: Alkali Scientific MPS002CP
AMPSO (3 ((1,1 Dimethyl 2 hydroxyethyl)amino) 2 hydroxypropanesulfonic acid) is a biochemical reagent used primarily as a buffering agent in laboratory settings. The 25g quantity is ideal for smaller experiments and offers sufficient material for preparing buffers for high-precision biochemical assays. AMPSO's ability to maintain a stable pH in various experimental conditions makes it essential for reactions where pH control is crucial. It is commonly used in molecular biology, protein chemistry, and enzymatic assays where pH fluctuations could interfere with the accuracy of results. The small volume is convenient for labs with limited storage space or for experiments with smaller sample sizes.