Learn More
PACIFIC BIOSCIENCES OF CALIFORNIA INC Pacific Biosciences AMPure PB Bead Size Selection Kit, Contaminant Removal, <5 kb DNA Depletion
Supplier:  PACIFIC BIOSCIENCES OF CALIFORNIA INC 102182500
Encompass
The AMPure PB bead size selection kit is essential for DNA library preparation, enabling diluted bead cleanup and precise size selection. This kit effectively removes contaminants and progressively depletes DNA fragments shorter than 5 kb from samples.
  • Used in procedures for preparing whole genome and metagenome libraries.
  • Effectively removes contaminants from DNA libraries.
  • Progressively depletes DNA fragments shorter than 5 kb.
  • Size selection performance is sensitive to bead concentrations, requiring accurate pipetting.
  • A 35% v/v dilution can be prepared with 1.75 mL of resuspended beads and 3.25 mL of elution buffer.
  • Diluted solution can be stored at 4°C for up to 30 days.
  • For each sample, 3.1X v/v (155 μL) of the 35% beads are added.
Catalog No. NC3977114