Learn More
Selleck Chemical LLC Amredobresib E4609-100MG
SDP
Supplier:  Selleck Chemical LLC E4609100MG
Encompass_Preferred
Amredobresib (BI894999) is an oral inhibitor of BET which acts as an acetyl-lysine mimic It prevents the bromodomains BRD4-BD1 and BRD4-BD2 from binding to acetylated histones with an IC50 of 5 nM for BRD4-BD1 and 41 nM for BRD4-BD2
Catalog No. 50-000-39532