Learn More
Enterprise Technology Solutions Amrep, Inc Fragrance Refill, f/Metered Disp, 6.6oz, Caribbean Waters
SDP
Supplier:  Enterprise Technology Solutions TMS1042756
Encompass_Preferred
Elevate your customer or visitor experience with this innovative, Premium Metered 30-Day Air Freshener - Caribbean Waters. Perfect for your odor control program, fragrance and odor counteractants in one can neutralize stubborn odors swiftly and provide lasting aromatherapy from the high-quality fragrance made with essential oils. Dual actuator system creates small micron dispersion, eliminating fallout. High fragrance load percentage ensures longest-lasting experience. The crystal-clear surf just offshore in the exotic Caribbean Islands sets the tone for this cool and soothing fragrance. Dual odor counteractants target specific malodor-causing particles. Use this refill in TimeMist full-sized, metered fragrance dispensers (not included) for 30 days of continuous use when set at 15-minute intervals.
Catalog No. 50-007-90460