Learn More
Apexbio Technology LLC Amrinone 60719-84-8 250mg
SDP
Supplier:  Apexbio Technology LLC A4663250
Encompass_Preferred
Amrinone (CAS 60719-84-8) is a small molecule inhibitor of type 3 phosphodiesterase (PDE3) enzymes By blocking PDE3 activity amrinone increases intracellular cyclic adenosine monophosphate (cAMP) levels leading to enhanced cardiac contractility and vasodilation This mechanism has made amrinone a valuable tool for investigating cardiac function and vascular smooth muscle regulation in preclinical models It is frequently utilized in research focused on the pathophysiology and treatment of congestive heart failure as well as studies exploring intracellular signaling pathways involving cAMP
Catalog No. 50-000-93785